Deb USA Universal Protect Pre-work Skin Cream


Overview
Brand: Deb USA

Manufacturer Part Number: UPW1L

X6 Skin cream, Deb Universal Protect, pre-work,

UNSPSC: 53131613

Code: 04

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 12839247

Quantity Price
1 £ 177.0 / Pack of 6
Estimated Shipment
07-03-2017
Add to basket


Description and Specification

Specification

Size (Metric) 999.88mL
Packaging Cartridge
Quantity 6 Case
Color Pale yellow
Composition Water, Kaolin, Cetearyl Alcohol, Dicaprylyl Carbonate, Glycerin, Xanthan Gum, Phenoxyethanol, Sodium Cetearyl Sulfate, Sodium Lauryl Sulfate, Methylparaben, 2-Bromo-2-Nitropropane-1,3-diol, Butylparaben, Ethylparaben, Isobutylparaben, Propylparaben, Yellow 5, Yellow 6
For Use With (Application) Helps protect the skin against contact with various workplace contaminants and working environments such as wet or dry condition
Form Cream
Shelf Life 24 Month(s)
Product Type Universal Protect Skin Cream
pH Range 6 to7.5