Deb USA Pure RESTORE™ Conditioning Cream


Overview
Brand: Deb USA

Manufacturer Part Number: RES1L

UNSPSC: 53131613

Code: 04

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 12829257

Quantity Price
1 £ 138.0 / Pack of 6
Estimated Shipment
11-05-2017
Add to basket


Description and Specification

Specification

Form Cream
Product Type Pure Restore Skin Conditioning Cream
Color White
Size (Metric) 999.88mL
Shelf Life 24 Month(s)
Packaging Cartridge
Quantity 6 Case
pH Range 6 to7.5
For Use With (Application) For use at the end of each work period to moisturize the skin while natural oils are being replenished, suitable for use on dry, sore, chapped or normal skin
Composition Water, Dicaprylyl Ether, Cetearyl Alcohol, Acrylates Copolymer, Glycerin, Phenoxyethanol, Butyrospermum Parkii (Shea) Butter, Ceteareth-20, Oleth-12, Aminomethyl Propanol, Titanium Dioxide, Allantoin, Methylparaben, Tetrasodium Iminodisuccinate , Butylparaben, Ethylparaben, Isobutylparaben, Propylparaben