Deb USA FOAM WASH Washroom Cleansers

Pleasantly fragranced, rich and mild foam soap with added moisturizers

Overview
Brand: Deb USA

Manufacturer Part Number: CLR1L

UNSPSC: 53131626

Code: 04

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 12879247

Quantity Price
1 £ 80.85 / Pack of 6
Estimated Shipment
09-05-2017
Add to basket


Description and Specification

Specification

Product Type Foam Wash Cleanser
Solution Type Clear FOAM WASH; 1L Cartridge
Color Clear
Shelf Life 2 Year(s)
Size (Metric) 1L
Format Liquid
Packaging Cartridge
Quantity 6 Case
Composition Water, Sodium Laureth Sulfate, Propylene Glycol, Aloe Barbadensis Leaf Juice, Cocamidopropyl Betaine, PEG-75 Lanolin, Allantoin, Hydroxypropyl Guar Hydroxypropyltrimonium Chloride, Benzyl Alcohol, Citric Acid, Methylchloroisothiazolinone, Methylisothiazolinone
pH Range 5 to 6
For Use With (Application) For general wash cleanse room Use
Certifications/Compliance Green Seal, Federal Food, Drug, and Cosmetic Act, NSF E1-rated