missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR1 Mouse anti-Human, Clone: 290420, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | DDR1 |
---|---|
Clone | 290420 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human DDR1 Asp19-Thr416 Accession # Q08345 |
Gene ID (Entrez) | 780 |
Gene Accession No. | Q08345 |
Description
DDR1 Monoclonal specifically detects DDR1 in Human samples. It is validated for Immunoprecipitation.Specifications
DDR1 | |
Mouse myeloma cell line NS0-derived recombinant human DDR1 Asp19-Thr416 Accession # Q08345 | |
Q08345 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
290420 | |
780 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human DDR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human DDR2. | |
Unconjugated | |
Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only