missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ DCX Recombinant Human Protein, N-GST Tag

Expressed from DNA sequence encoding human DCX N-terminal fragment (Ala 45-Val 150) fused to GST tag at N-terminus

Brand:  Sino Biological™ 13498H09E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15226315

  • £232.15 / 50µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 1mM DTT, 10% glycerol, pH 7.5
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
1641
≥90% by SDS-PAGE
Human
<1 EU/μg
DCX
Cell Structure/Organelle Proteins
DCX Recombinant Human Protein
Lyophilized
39.4kd
50μg
Truncated, Recombinant
Doublecortin
Assay Standard
E.coli
GST Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.