missing translation for 'onlineSavingsMsg'
Learn More

DCPIB, Tocris Bioscience™

Blocker of VRAC/ICl, swell

Brand:  Tocris (Bio-Techne) 1540/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13449633

  • £149.48 / 10mg

Estimated Shipment
15-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Volume-regulated anion channel (VRAC) blocker (IC50 ∽ 2 μM in rat pancreatic β-cells). Also blocks ICl,swell in various cardiovascular tissues (IC50 = 4.1 μM in CPAE cells). Inhibits glucose-stimulated insulin secretion in intact β-cells via VSAC inhibition and indirect KATP channel activation. Reverses cell swelling-induced action potential duration shortening in atrial myocytes and inhibits astroglial swelling in vitro. Also activates TREK1 and TRAAK K+ channels and inhibits TRESK, TASK1 and TASK3 K+ channels at 10 μM, in vitro.
Specifications

Chemical Identifiers

82749-70-0
427.362
dcpib, 4-2-butyl-6,7-dichlor-2-cyclopentyl-indan-1-on-5-yl oxybutyric acid, d0j3qc, 4-2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-3h-inden-5-yl oxy butanoic acid, 4-2-butyl-6,7-dichloro-2-cyclopentylindan-1-on-5-yl oxybutyric acid, 4-2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-2,3-dihydro-1h-inden-5-yl oxy butanoic acid, 4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid, + 4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid, -4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid
4-[(2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-3H-inden-5-yl)oxy]butanoic acid
C22H28Cl2O4
KHKGTPJPBOQECW-UHFFFAOYSA-N
10071166
CCCCC1(CC2=CC(=C(C(=C2C1=O)Cl)Cl)OCCCC(=O)O)C3CCCC3

Specifications

DCPIB
82749-70-0
dcpib, 4-2-butyl-6,7-dichlor-2-cyclopentyl-indan-1-on-5-yl oxybutyric acid, d0j3qc, 4-2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-3h-inden-5-yl oxy butanoic acid, 4-2-butyl-6,7-dichloro-2-cyclopentylindan-1-on-5-yl oxybutyric acid, 4-2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-2,3-dihydro-1h-inden-5-yl oxy butanoic acid, 4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid, + 4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid, -4-2-butyl-6,7-dichloro-2-cyclopentyl-2,3-dihydro-1-oxo-1h-inden-5-yl oxy butanoic acid
CCCCC1(CC2=CC(=C(C(=C2C1=O)Cl)Cl)OCCCC(=O)O)C3CCCC3
427.362
427.37
10mg
C22H28Cl2O4
KHKGTPJPBOQECW-UHFFFAOYSA-N
4-[(2-butyl-6,7-dichloro-2-cyclopentyl-1-oxo-3H-inden-5-yl)oxy]butanoic acid
10071166
>98%
Videos
Documents