Learn More
DCHS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562158
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TVSVLVTGED EYDPVFLAPA FHFQVPEGAR RGHSLGHVQA TDEDGGADGL VLYSLATSSP YFGINQTTGA LYLRVDSRAP GSGTATSGGG GRTRREAPRE LRLEVIARGP LPGSRSATVP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
DCHS1 Polyclonal antibody specifically detects DCHS1 in Human samples. It is validated for Immunohistochemistry.Specifications
DCHS1 | |
Polyclonal | |
Unconjugated | |
DCHS1 | |
Cadherin-19; cadherin-25; cadherin-related family member 6; CDH19; CDH25; CDHR6; dachsous cadherin-related 1; DCHS1; FIB1; fibroblast cadherin 1; fibroblast cadherin FIB1; Fibroblast cadherin-1; KIAA1773; MVP2; PCDH16; Protein dachsous homolog 1; protocadherin-16; VMLDS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8642 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JQ0 | |
DCHS1 | |
Recombinant protein corresponding to Human DCHS1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only