Learn More
DCB, Tocris Bioscience™
Allosteric modulator at mGlu5
Brand:  Tocris (Bio-Techne) 195250
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13460284
  • £257.41 / 50mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Allosteric ligand for the metabotropic glutamate receptor mGlu5; displays neutral modulation. Does not affect agonist-stimulated mGlu5 responses, but blocks regulation of the receptor by other allosteric modulators such as DFB (Cat. No. 1625) and DMeOB (Cat. No. 1953).
Specifications
Chemical Identifiers
6971-97-7
277.148
3,3'-dichlorobenzylideneazine, benzaldehyde, 3-chlorophenyl methylene hydrazone
(E)-1-(3-chlorophenyl)-N-[(Z)-(3-chlorophenyl)methylideneamino]methanimine
C14H10Cl2N2
XMOVWXSCYLINBJ-BUOZRGFLSA-N
6861558
C1=CC(=CC(=C1)Cl)C=NN=CC2=CC(=CC=C2)Cl
Specifications
DCB
6971-97-7
3,3'-dichlorobenzylideneazine, benzaldehyde, 3-chlorophenyl methylene hydrazone
C1=CC(=CC(=C1)Cl)C=NN=CC2=CC(=CC=C2)Cl
277.148
277.15
50mg
C14H10Cl2N2
XMOVWXSCYLINBJ-BUOZRGFLSA-N
(E)-1-(3-chlorophenyl)-N-[(Z)-(3-chlorophenyl)methylideneamino]methanimine
6861558
>98%
Videos