missing translation for 'onlineSavingsMsg'
Learn More

DAU 5884 hydrochloride, Tocris Bioscience™

M3 receptor antagonist

Brand:  Tocris (Bio-Techne) 2096/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13460464

  • £117.45 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Selective muscarinic M3 receptor antagonist. In bovine tracheal smooth muscle, inhibits methacholine-dependent effects on cell proliferation and muscle contractility.
Specifications

Chemical Identifiers

131780-48-8
351.831
dau 5884 hydrochloride, 8-methyl-8-azabicyclo-3-endo 3.2.1 oct-3-yl-1,4-dihydro-2-oxo-3 2h-quinazolinecarboxylic acid ester hydrochloride, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2-oxo-1,4-dihydroquinazoline-3-carboxylate hydrochloride, 1r,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2-oxo-1,4-dihydroquinazoline-3-carboxylate hydrochloride, 1r,5s-rel-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2-oxo-1,2-dihydroquinazoline-3 4h-carboxylate hydrochloride, 8-methyl-8-azabicyclo-3-endo 3.2.1 oct-3-yl-1,4-dihydro-2-oxo-3 2h-quinazolinecarboxylic acid ester hydrochloride, 8-methyl-8-azabicyclo-3-endo 3.2.1 oct-3-yl-1,4-dihydro-2-oxo-3 2h-quinazolinecarboxylic acid ester hydrochloride
[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 2-oxo-1,4-dihydroquinazoline-3-carboxylate;hydrochloride
C17H22ClN3O3
FDERDDSQHZRNGC-LIWIJTDLSA-N
16759154
CN1C2CCC1CC(C2)OC(=O)N3CC4=CC=CC=C4NC3=O.Cl

Specifications

DAU 5884 hydrochloride
131780-48-8
dau 5884 hydrochloride, 8-methyl-8-azabicyclo-3-endo 3.2.1 oct-3-yl-1,4-dihydro-2-oxo-3 2h-quinazolinecarboxylic acid ester hydrochloride, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2-oxo-1,4-dihydroquinazoline-3-carboxylate hydrochloride, 1r,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2-oxo-1,4-dihydroquinazoline-3-carboxylate hydrochloride, 1r,5s-rel-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2-oxo-1,2-dihydroquinazoline-3 4h-carboxylate hydrochloride, 8-methyl-8-azabicyclo-3-endo 3.2.1 oct-3-yl-1,4-dihydro-2-oxo-3 2h-quinazolinecarboxylic acid ester hydrochloride, 8-methyl-8-azabicyclo-3-endo 3.2.1 oct-3-yl-1,4-dihydro-2-oxo-3 2h-quinazolinecarboxylic acid ester hydrochloride
CN1C2CCC1CC(C2)OC(=O)N3CC4=CC=CC=C4NC3=O.Cl
351.831
351.83
10mg
C17H22ClN3O3
FDERDDSQHZRNGC-LIWIJTDLSA-N
[(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 2-oxo-1,4-dihydroquinazoline-3-carboxylate;hydrochloride
16759154
>99%
Videos