Learn More
DARS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554210
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LHVQKIYVIS LAEPRLPLQL DDAVRPEAEG EEEGRATVNQ DTRLDNRVID LRTSTSQAVF RLQSGICHLF RETLINKGFV EIQTPKIISA ASEGGANVFT VSYFKNNAYL AQSPQLYKQM CICAD Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.
DARS Polyclonal antibody specifically detects DARS in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DARS | |
Polyclonal | |
Unconjugated | |
DARS1 | |
5730439G15Rik; aspartate tRNA ligase 1, cytoplasmic; aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; aspRS; Cell proliferation-inducing gene 40 protein; Dars; DRS1; HBSL; PIG40; testicular tissue protein Li 192 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P14868 | |
DARS1 | |
Recombinant protein corresponding to Human DARS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only