missing translation for 'onlineSavingsMsg'
Learn More

DAPT, Tocris Bioscience™

γ-secretase inhibitor; induces neuronal differentiation

Brand:  Tocris (Bio-Techne) 263410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13451614

  • £219.03 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of γ-secretase; causes a reduction in Aβ40 and Aβ42 levels in human primary neuronal cultures (IC50 values are 115 and 200 nM for total Aβ and Aβ42 respectively) and in brain extract, cerebrospinal fluid and plasma in vivo. Does not affect APPα and APPβ levels. Blocks Notch signaling in hybrid human-mouse fetal thymus organ culture (FTOC). Activity causes ESCs to commit to neuronal differentiation.

DAPT synthesized to cGMP guidelines also available.

Specifications

Chemical Identifiers

208255-80-5
432.468
dapt, dapt gsi-ix, gamma-secretase inhibitor ix, gsi-ix, insolution gamma-secretase inhibitor ix, s-tert-butyl 2-s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, n-n-3,5-difluorophenacetyl-l-alanyl-s-phenylglycine t-butyl ester, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetyl amino propanoyl amino-2-phenylacetate, gsiix
CHEBI:86193
CC(C(=O)NC(C1=CC=CC=C1)C(=O)OC(C)(C)C)NC(=O)CC2=CC(=CC(=C2)F)F
C23H26F2N2O4
DWJXYEABWRJFSP-XOBRGWDASA-N
5311272
tert-butyl (2S)-2-[[(2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]propanoyl]amino]-2-phenylacetate

Specifications

10mg
208255-80-5
dapt, dapt gsi-ix, gamma-secretase inhibitor ix, gsi-ix, insolution gamma-secretase inhibitor ix, s-tert-butyl 2-s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, n-n-3,5-difluorophenacetyl-l-alanyl-s-phenylglycine t-butyl ester, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetyl amino propanoyl amino-2-phenylacetate, gsiix
CC(C(=O)NC(C1=CC=CC=C1)C(=O)OC(C)(C)C)NC(=O)CC2=CC(=CC(=C2)F)F
432.468
CHEBI:86193
>99%
DAPT
C23H26F2N2O4
DWJXYEABWRJFSP-XOBRGWDASA-N
tert-butyl (2S)-2-[[(2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]propanoyl]amino]-2-phenylacetate
5311272
432.46
Videos
Documents