missing translation for 'onlineSavingsMsg'
Learn More

DAPT, Tocris Bioscience™

γ-secretase inhibitor

Brand:  Tocris (Bio-Techne) 263450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461614

  • £710.36 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Inhibitor of γ-secretase; causes a reduction in Aβ40 and Aβ42 levels in human primary neuronal cultures (IC50 values are 115 and 200 nM for total Aβ and Aβ42 respectively) and in brain extract, cerebrospinal fluid and plasma in vivo. Does not affect APPα and APPβ levels. Blocks Notch signaling in hybrid human-mouse fetal thymus organ culture (FTOC). Activity causes ESCs to commit to neuronal differentiation.
Specifications

Chemical Identifiers

208255-80-5
432.468
dapt, dapt gsi-ix, gamma-secretase inhibitor ix, gsi-ix, insolution gamma-secretase inhibitor ix, s-tert-butyl 2-s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, n-n-3,5-difluorophenacetyl-l-alanyl-s-phenylglycine t-butyl ester, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetyl amino propanoyl amino-2-phenylacetate, gsiix
CHEBI:86193
CC(C(=O)NC(C1=CC=CC=C1)C(=O)OC(C)(C)C)NC(=O)CC2=CC(=CC(=C2)F)F
C23H26F2N2O4
DWJXYEABWRJFSP-XOBRGWDASA-N
5311272
tert-butyl (2S)-2-[[(2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]propanoyl]amino]-2-phenylacetate

Specifications

DAPT
208255-80-5
dapt, dapt gsi-ix, gamma-secretase inhibitor ix, gsi-ix, insolution gamma-secretase inhibitor ix, s-tert-butyl 2-s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, n-n-3,5-difluorophenacetyl-l-alanyl-s-phenylglycine t-butyl ester, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetyl amino propanoyl amino-2-phenylacetate, gsiix
CC(C(=O)NC(C1=CC=CC=C1)C(=O)OC(C)(C)C)NC(=O)CC2=CC(=CC(=C2)F)F
432.468
CHEBI:86193
>99%
50mg
C23H26F2N2O4
DWJXYEABWRJFSP-XOBRGWDASA-N
tert-butyl (2S)-2-[[(2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]propanoyl]amino]-2-phenylacetate
5311272
432.46
Videos