missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAPK2 Recombinant Human Protein
DAPK2 is a calcium/calmodulin-dependent kinase.
£1602.00 - £6900.00
Specifications
Accession Number | NP_055141 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 300mM NaCl, 0.5mM EDTA, 0.05% Brij-35, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 47.9kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478453
|
Invitrogen™
PR5688A |
100μg |
£1602.00
100µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10168912
|
Invitrogen™
PV3994 |
1mg |
£6900.00
1mg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DAPK2 acts as a positive regulator of apoptosis and is expressed in many tissue types.
Specifications
NP_055141 | |
50mM Tris-HCl, pH 7.5, 300mM NaCl, 0.5mM EDTA, 0.05% Brij-35, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
DAPK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
47.9kDa | |
RUO - research use only | |
DAPK2, DRP-1 | |
Human | |
His Tag | |
Frozen |