missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAN Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | DAN/NBL1 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human DAN Ala17-Asp180 (Ala36Arg) Accession # Q5U0N4 |
Gene ID (Entrez) | 4681 |
Gene Accession No. | Q5U0N4 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
DAN/NBL1 Polyclonal specifically detects DAN/NBL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
DAN/NBL1 | |
4681 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human DAN in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) DAN is observed and less than 1% cross-reactivity with recombinant chicken Caronte, rmGremlin, and recombinant human Cerberus 1 is observed. | |
Unconjugated | |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human DAN Ala17-Asp180 (Ala36Arg) Accession # Q5U0N4 | |
Q5U0N4 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only