missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549237
Additional Details : Weight : 0.01000kg
Description
Predicted to react with rat based on sequence homology.
DAB2 Polyclonal antibody specifically detects DAB2 in Mouse samples. It is validated for Western Blot.Specifications
DAB2 | |
Polyclonal | |
Unconjugated | |
DAB2 | |
5730435J12Rik; AA960054; Adaptor molecule disabled-2; AI957090; C9; D15Wsu122e; D630005B22Rik; Dab, mitogen-responsive phosphoprotein, homolog 2; Dab2; DAB2, clathrin adaptor protein; differentially expressed in ovarian carcinoma 2; Differentially-expressed protein 2; disabled 2, mitogen-responsive phosphoprotein; disabled homolog 2; disabled homolog 2 mitogen-responsive phosphoprotein; disabled homolog 2, mitogen-responsive phosphoprotein; Doc2; Doc-2; DOC-2 p82 isoform; FLJ26626; Mitogen-responsive phosphoprotein; p96 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
13132 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P98078 | |
DAB2 | |
KLH conjugated synthetic peptide between 624-650 amino acids from the C-terminal region of mouse Dab2. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |