Learn More
Thermo Scientific Chemicals D-(+)-Xylose, 98+%
Description
D-(+)-Xylose is used to prepare furfural by acid catalyzed degradation reaction, which acts as a solvent as well as a precursor to synthetic polymers. It is a useful animal medicine involved in the treatment of malabsorption. It finds application in the production of sugar substitute xylitol by subjecting xylose to calyticl hydrogenation process. It is found that it undergoes an atmospheric-pressure enzyme (polyphosphate xylulokinase) driven process to get hydrogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-(+)-Xylose is used to prepare furfural by acid catalyzed degradation reaction, which acts as a solvent as well as a precursor to synthetic polymers. It is a useful animal medicine involved in the treatment of malabsorption. It finds application in the production of sugar substitute xylitol by subjecting xylose to calyticl hydrogenation process. It is found that it undergoes an atmospheric-pressure enzyme (polyphosphate xylulokinase) driven process to get hydrogen.
Solubility
Soluble in water.
Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 147°C to 151°C |
Density | 1.525 |
Flash Point | > 100°C (212°F) |
Assay Percent Range | >98% |
Quantity | 100 g |
Beilstein | 1562108 |
Sensitivity | Hygroscopic |
Merck Index | 14,10087 |
Solubility Information | Soluble in water. |
Optical Rotation | +19° (c=10 in water + NH4OH) |
Show More |
RUO – Research Use Only