missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Tryptophan, 99%, ACROS Organics™

Brand:  Acros Organics 168635000

UNSPSC : 12352209

Code : 29

Additional Details : CAS Number : 153-94-6 Weight : 0.55000kg

Quantity 500g

 View more versions of this product

Product Code. 11966611

  • £771.60 / 500g


Estimated Shipment
21-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

D(+)-Tryptophan
99%
204.23
≥98.5% (total base)
C11H12N2O2

Specifications

D(+)-Tryptophan
(1% aq. soln.) Clear colorless to light yellow
≥98.5% (total base)
Authentic
204.23
+29.5° to +33.5° (20°C, 589nm) (c=1, H2O)
White to slightly yellow
153-94-6
Powder
99%
C11H12N2O2
0.5% max.
+ 31.50 (24.00°C c=1,H2O)
500g

Safety and Handling

Product Identifier
  • D(+)-Tryptophan

Documentation

Technical Documents