missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, Thermo Scientific Chemicals

Catalog Number p-194788
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
147-71-7
C4H6O6
150.09
MFCD00004238
FEWJPZIEWOKRBE-UHFFFAOYNA-N
d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
439655
CHEBI:15672
OC(C(O)C(O)=O)C(O)=O
This item is not returnable. View return policy
147-71-7
C4H6O6
150.09
MFCD00004238
FEWJPZIEWOKRBE-UHFFFAOYNA-N
d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
439655
CHEBI:15672
OC(C(O)C(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 167°C to 171°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH(OH)CH(OH)CO2H
Beilstein 03, 520
Fieser 16,304
Merck Index 15, 9203
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid
Formula Weight 150.09
Percent Purity ≥98.5%
Quantity 25 g
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Packaging Glass Bottle
Physical Form Crystals or Crystalline Powder
Chemical Name or Material D(-)-Tartaric acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'