missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Tagatose, 99+%, ACROS Organics™

Brand:  Acros Organics 257890010

Code : 29

Additional Details : CAS Number : 87-81-0 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10398320

  • £75.90 / 1g


EU Stock 64
Add to basket

Specifications

Chemical Identifiers

D(-)-Tagatose
≥99.0%
C6H12O6
MFCD00134449
87-81-0
99+%
180.16
D-lyxo-Hexulose

Specifications

D(-)-Tagatose
(5% soln. in H2O) clear colorless to almost colorless
Crystalline Powder
99+%
180.16
MFCD00134449
15, 9161
Glass bottle
0.1% max.
White to off-white
87-81-0
Authentic
≥99.0%
C6H12O6
1.0% max.
01, 4414
D-lyxo-Hexulose
-5° to -7° (20°C, 589nm) (c=2, H2O)
- 6.00 (20.00°C c=5,water)
1g
Videos
SDS