missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Sorbitol, 98%
28.12 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
Description
D-sorbitol is added to food products as sweetener. It is also used in industrial and medical applications. Sorbitol often is applied in modern cosmetics as a humectant and thickener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 98°C to 100°C |
Assay Percent Range | 98% |
Quantity | 500 g |
Beilstein | 1721899 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water,methanol,acetone and DMF. |
Formula Weight | 182.17 |
Physical Form | Powder |
Chemical Name or Material | D-Sorbitol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'