missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Serine methyleester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 152.0°C to 161.0°C |
Color | Colorless to Yellow |
Assay Percent Range | 98% |
Infrared Spectrum | Authentic |
Linear Formula | HOCH2CH(NH2)CO2CH3·HCl |
Packaging | Glass Bottle |
Specific Rotation | −2.5° to −4.5° (20°C, 589nm) (C=4, methanol) |
Formula Weight | 155.58 |
Percent Purity | 98% |
Quantity | 5 g |
Show More |
Safety and Handling
Product Identifier
- D-Serine methyl ester hydrochloride
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'