missing translation for 'onlineSavingsMsg'
Learn More

D-Serine, 98%, Thermo Scientific Chemicals

Catalog Number 11921731
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
312-84-5
C3H7NO3
105.093
MFCD00004269
MTCFGRXMJLQNBG-UWTATZPHSA-N
d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
71077
CHEBI:16523
(2R)-2-amino-3-hydroxypropanoic acid
C(C(C(=O)O)N)O
This item is not returnable. View return policy
312-84-5
C3H7NO3
105.093
MFCD00004269
MTCFGRXMJLQNBG-UWTATZPHSA-N
d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
71077
CHEBI:16523
(2R)-2-amino-3-hydroxypropanoic acid
C(C(C(=O)O)N)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 220.0°C
Assay Percent Range 98%
Linear Formula HOCH2CH(NH2)CO2H
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water: 346g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene., soluble in mineral acids., insoluble in organic solvents.
Specific Rotation − 14.75
Formula Weight 105.09
Percent Purity 98%
Specific Rotation Condition −14.75° (20°C c=10 2 N HCl)
Quantity 500 g
Chemical Name or Material D-Serine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'