missing translation for 'onlineSavingsMsg'
Learn More

D-Proline ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7056147
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
131477-20-8
C7H14ClNO2
179.644
MFCD00191003
DUJGQVVONTYHLT-FYZOBXCZSA-N
r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl
56777374
ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CCCN1.Cl
This item is not returnable. View return policy
131477-20-8
C7H14ClNO2
179.644
MFCD00191003
DUJGQVVONTYHLT-FYZOBXCZSA-N
r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl
56777374
ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CCCN1.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Beilstein 7170607
Formula Weight 179.64
Percent Purity 98%
Quantity 5 g
Physical Form Liquid
Chemical Name or Material D-Proline ethyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'