missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ D-Proline ethyl ester hydrochloride, 98%

Brand:  Alfa Aesar™ H64910.22

Code : NEW

Additional Details : CAS Number : 131477-20-8 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15412329

  • £260.20 / 100g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

D-Proline ethyl ester hydrochloride
179.64
7170607
131477-20-8
MFCD00191003
Ethyl (R)-pyrrolidine-2-carboxylate hydrochloride

Specifications

D-Proline ethyl ester hydrochloride
Liquid
179.64
7170607
Pale yellow
131477-20-8
98%
MFCD00191003
Ethyl (R)-pyrrolidine-2-carboxylate hydrochloride
100g
Videos
SDS