missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Proline, ≥99%

Catalog Number p-3776639
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
344-25-2
C5H9NO2
115.132
MFCD00064317
ONIBWKKTOPOVIA-SCSAIBSYSA-N
d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
8988
CHEBI:16313
(2R)-pyrrolidine-2-carboxylic acid
C1CC(NC1)C(=O)O
This item is not returnable. View return policy
344-25-2
C5H9NO2
115.132
MFCD00064317
ONIBWKKTOPOVIA-SCSAIBSYSA-N
d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
8988
CHEBI:16313
(2R)-pyrrolidine-2-carboxylic acid
C1CC(NC1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown to White
Assay Percent Range 99+%
Infrared Spectrum Authentic
Beilstein 22, 2
Fieser 07,307; 09,393
Loss on Drying 1 % max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Glass Bottle
Specific Rotation +83.0° to +87.0° (20°C, 589nm) (c=4, water)
Formula Weight 115.13
Percent Purity 99.0 to 101.0% (Total base)
Sulfated Ash 0.1 % max.
Specific Rotation Condition +84.75 (20.00°C c=4,H2O)
Quantity 1 g
Physical Form Powder
Chemical Name or Material D(+)-Proline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'