Alfa Aesar™ D-Pipecolinic acid, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H63255.14

25GR D-Pipecolinic acid, 97%

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: CAS Number: 1723-00-8 Weight: 0.02500kg


Quantity 25g

Product Code. 15419858

Quantity Price
1 £ 74.84 / 25g
Estimated Shipment date
from Supplier 10-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material D-Pipecolinic acid
CAS 1723-00-8
Formula Weight 129.16
MDL Number MFCD00064346
Beilstein 81094
Synonym D-Homoproline

Specification

Formula Weight 129.16
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material D-Pipecolinic acid
Physical Form Solid
Assay 97%
MDL Number MFCD00064346
CAS 1723-00-8
Beilstein 81094
Synonym D-Homoproline
Health Hazard 1 Warning
Melting Point 277°C (dec.)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Safety and Handling

Warning

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.