missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 11935391
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
13033-84-6
C10H14ClNO2
215.68
MFCD00066112
SWVMLNPDTIFDDY-SBSPUUFOSA-N
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
11481330
methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
Cl.COC(=O)[C@H](N)CC1=CC=CC=C1
This item is not returnable. View return policy
13033-84-6
C10H14ClNO2
215.68
MFCD00066112
SWVMLNPDTIFDDY-SBSPUUFOSA-N
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
11481330
methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
Cl.COC(=O)[C@H](N)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 157°C to 159°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Specific Rotation -35.76
Formula Weight 215.68
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material D-Phenylalanine methyl ester hydrochloride
Product Identifier
  • D-Phenylalanine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'