missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
27.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 157°C to 159°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 5 g |
Specific Rotation | -35.76 |
Formula Weight | 215.68 |
Percent Purity | 98% |
Physical Form | Crystalline Powder or Powder |
Chemical Name or Material | D-Phenylalanine methyl ester hydrochloride |
Safety and Handling
Product Identifier
- D-Phenylalanine methyl ester hydrochloride
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'