D-Phenylalanine methyleester hydrochloride, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 442620250

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 13033-84-6 Weight: 0.07500kg


Packaging Glass bottle
Quantity 25g

Product Code. 11955391

Quantity Price
1 £ 99.3 / 25g
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material D-Phenylalanine methyleester hydrochloride
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H13NO2·HCl
Formula Weight 215.68

Specification

Formula Weight 215.68
Assay ≥97.5%
Physical Form Powder or Crystalline Powder
Quantity 25g
Color White to off-white
Specific Rotation -32° to -38° (20°C, 589nm) (c=1 in EtOH)
CAS 13033-84-6
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H13NO2·HCl
Chemical Name or Material D-Phenylalanine methyleester hydrochloride
Documentation

Technical documents