missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Phenylalanine, 99%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.
Solubility
Soluble in water (30 mg/ml). Slightly soluble in methanol, ethanol, DMSO.
Notes
Store at room temperature. Incompatible with oxidizing agents.
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.
Solubility
Soluble in water (30 mg/ml). Slightly soluble in methanol, ethanol, DMSO.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | ∽273°C to 276°C (decomposition) |
Beilstein | 2804068 |
Merck Index | 14,7271 |
Solubility Information | Soluble in water (30mg/ml). Slightly soluble in methanol,ethanol,DMSO. |
Formula Weight | 165.19 |
Percent Purity | 99% |
Quantity | 5 g |
Optical Rotation | +34° (c=2 in water) |
Chemical Name or Material | D-Phenylalanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'