missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Penicillamine, 99%, specified according to Ph.Eur. and USP

Catalog Number 10061450
41.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
52-67-5
C5H11NO2S
149.21
MFCD00064302
VVNCNSJFMMFHPL-VKHMYHEASA-N
d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
5852
CHEBI:7959
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
CC(C)(C(C(=O)O)N)S
This item is not returnable. View return policy
52-67-5
C5H11NO2S
149.21
MFCD00064302
VVNCNSJFMMFHPL-VKHMYHEASA-N
d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
5852
CHEBI:7959
(2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
CC(C)(C(C(=O)O)N)S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 0.07 max. (c=0.2, H2O, 1 cm cell) at 268nm
Melting Point 212.0°C
Color White
pH 4.5 to 5.5 (1% soln.)
Ignition Residue 0.1% max
Loss on Drying 0.5% max. (1 g, 60°C, 670 Pa) (P2O5)
Infrared Spectrum Authentic
Assay Percent Range 97.0 to 102% (USP)
Packaging Glass bottle
Linear Formula (CH3)2C(SH)CH(NH2)CO2H
Quantity 5 g
Merck Index 15, 7197
Specific Rotation Condition −65° (20°C c=5,1M NaOH,on dry)
Specific Rotation − 65.00
Solubility Information Solubility in water: 11.1% (20°C). Other solubilities: slightly soluble in ethanol,insoluble in ether,benzene,acetone,carbon tetra,chloride
Formula Weight 149.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D(-)-Penicillamine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • D(-)-Penicillamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'