missing translation for 'onlineSavingsMsg'
Learn More

D-Myo-inositol, 99.26%, For HPLC analysis, MP Biomedicals™

Catalog Number p-4405728
Click to view available options
Quantity:
50 g
100 g
500 g
This item is not returnable. View return policy
87-89-8
C6H12O6
180.156
CDAISMWEOUEBRE-UHFFFAOYSA-N
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
892
CHEBI:24848
cyclohexane-1,2,3,4,5,6-hexol
C1(C(C(C(C(C1O)O)O)O)O)O
This item is not returnable. View return policy
87-89-8
C6H12O6
180.156
CDAISMWEOUEBRE-UHFFFAOYSA-N
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
892
CHEBI:24848
cyclohexane-1,2,3,4,5,6-hexol
C1(C(C(C(C(C1O)O)O)O)O)O
Color White
Density 6.2g/mL
Assay Percent Range 99.26%
Quantity 50 g
Formula Weight 180.2
Physical Form Powder
Chemical Name or Material D-Myo-inositol, For HPLC analysis
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'