missing translation for 'onlineSavingsMsg'
Learn More

α-D-Methylglucoside, 99%, ACROS Organics™

Brand:  Acros Organics 173840025

Code : 29

Additional Details : CAS Number : 97-30-3 Weight : 2.55000kg

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10579063

  • £227.50 / 2.50kg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

α-D-Methylglucoside
97-30-3
100.0
99%
194.18
Methyl-α-D-glucopyranoside
99%
98.5
99%
C7H14O6
MFCD00064086

Specifications

α-D-Methylglucoside
200.0°C (0.2 mmHg)
169.0° to 171.0°C
99%
98.5
Authentic
Plastic bottle
Fine Crystalline Powder
99%
194.18
31, 179
Methyl-α-D-glucopyranoside
+158.50° (20°C c=10,water)
White
2.5kg
97-30-3
100.0
0.1% max. (60 to 70°C)
+ 158.50
99%
C7H14O6
MFCD00064086
15, 6152
Videos
SDS