missing translation for 'onlineSavingsMsg'
Learn More

D-Methionine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 15420109
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
69630-60-0
C6H14ClNO2S
199.69
MFCD00070385
MEVUPUNLVKELNV-NUBCRITNSA-N
h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride
12888616
methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Cl.COC(=O)[C@H](N)CCSC
This item is not returnable. View return policy
69630-60-0
C6H14ClNO2S
199.69
MFCD00070385
MEVUPUNLVKELNV-NUBCRITNSA-N
h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride
12888616
methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Cl.COC(=O)[C@H](N)CCSC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 199.7
Percent Purity 98%
Quantity 25 g
Physical Form Solid
Chemical Name or Material D-Methionine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'