missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ D-Leucine benzyl ester p-toluenesulfonate, 98%

Brand:  Alfa Aesar™ H63815.06

Code : NEW

Additional Details : CAS Number : 17664-93-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15490709

  • £32.01 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

D-Leucine benzyl ester p-toluenesulfonate
393.5
3566840
17664-93-6
MFCD00066117
H-D-Leu-OBzl.Tos-OH

Specifications

D-Leucine benzyl ester p-toluenesulfonate
Powder
393.5
3566840
White
5g
17664-93-6
98%
MFCD00066117
H-D-Leu-OBzl.Tos-OH
153° to 160°C
Hygroscopic
Videos
SDS