missing translation for 'onlineSavingsMsg'
Learn More

β-D-Lactose, ca 80% β and 20% α, ACROS Organics™

Brand:  Acros Organics 412985000

Code : 29

Additional Details : CAS Number : 5965-66-2 Weight : 0.55000kg

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10528132

  • £27.90 / 500g


EU Stock 6
Add to basket

Specifications

Chemical Identifiers

β-D-Lactose
C12H22O11
MFCD00064521
≥78% (beta-lactose), 22% max. (alpha-lactose)
342.29

Specifications

Passes Test (per Eur. Pharm.)
5965-66-2
Passes Test (per Eur. Pharm.)
≥78% (beta-lactose), 22% max. (alpha-lactose)
5ppm max.
MFCD00064521
0.5% max. (1 g, 80°C, 2 hrs)
Plastic bottle
0.1% max.
+55.15° (20°C c=10,water,NH3)
500g
β-D-Lactose
0.4mL 0.1N NaOH/6g max.
Crystalline Powder
C12H22O11
342.29
Authentic
Passes Test (per Eur. Pharm.)
On anhydrous basis: +54.4° to +55.9° (20°C, 589nm) (c=10, H2O/NH3)
1.0% max. (K.F.)
White or almost white
Videos
SDS