missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

Catalog Number 10373242
55.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


73.11 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
500 g
1 kg
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
50-99-7
C6H12O6
180.156
148912
GZCGUPFRVQAUEE-SLPGGIOYSA-N
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
107526
CHEBI:42758
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
50-99-7
C6H12O6
180.156
148912
GZCGUPFRVQAUEE-SLPGGIOYSA-N
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
107526
CHEBI:42758
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O

C6H12O6, CAS Number-50-99-7, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, dextrose solution, aldehydo-d-glucose, dextrose, anhydrous, anhydrous dextrose, d-glucose in linear form, d-glucose, anhydrous, d +-glucose, glucose, anhydrous, glucosteril, 5kg, CHEBI:42758, White, 180.16g/mol

TRUSTED_SUSTAINABILITY
Melting Point 146°C
Color White
pH 6 to 7
Packaging Plastic powder jar
Quantity 1 kg
Formula Weight 180.16g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'