missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Glucose, 1M aq. soln., sterile-filtered, Thermo Scientific Chemicals
Click to view available options
Quantity:
125 mL
250 mL
Description
D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.
Solubility
Not miscible or difficult to mix with water. Soluble in methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
D-(+)-Glucose used as a ideal in culture media component. Also used in foods and medicines and as a source of certain amino acids by fermentation.
Solubility
Not miscible or difficult to mix with water. Soluble in methanol.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Quantity | 125 mL |
Solubility Information | Not miscible or difficult to mix with water. Soluble in methanol. |
Formula Weight | 180.16 |
Concentration or Composition (by Analyte or Components) | 1M aq. soln. |
Physical Form | Liquid |
Chemical Name or Material | D-(+)-Glucose, sterile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'