missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucose, Merck Millipore

Anhydrous for biochemistry (Reag. Ph Eur)

Brand:  Merck Millipore 1083370250

Code : NEW

Additional Details : CAS Number : 50-99-7 Weight : 0.25000kg

Quantity 250 g

 View more versions of this product

Product Code. 10122730

Quantity Price
1 £38.48 / 250g
10 £33.04 / 250g
Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

  • Application:
    • D(+)-Glucose anhydrous for biochemistry Reag. Ph Eur. CAS 50-99-7, molar mass 180.16 g/mol
  • Analysis note:
    • Assay (calc. on anhydrous substance)97.5 - 102.0 %
    • Identity (specific rotation) passes test
    • Identity (HPLC) passes test
    • Identity (water) passes test
    • Appearance of solution passes test
    • Conductivity ≤ 20 µS/cm
    • Related substances (sum of impurities A and B) ≤ 0.4 %
    • Related substances (impurity C) ≤ 0.2 %
    • Related substances (impurity D) ≤ 0.15 %
    • Related substances (unspecified impurities) ≤ 0.10 %
    • Related substances (Sum of all impurities) ≤ 0.5 %
    • Dextrin passes test
    • Soluble starch, sulfite ≤ 15 ppm
    • Residual solvents (ICH Q3C) excluded by the manufacturing process
    • Water (according to Karl Fischer) ≤ 1.0 %
    • Corresponds to Reag. Ph Eur.
Specifications

Chemical Identifiers

50-99-7
1 S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11 H,1H2/t2-,3-,4+,5-,6+/m1/s1
C6H12O6
180.16
470 g/L
25800 mg/kg LD50, oral (Rat)
WQZGKKKJIJFFOK-UHFFFAOYNA-N
MFCD00063774
OCC1OC(O)C(O)C(O)C1O
Dextrose

Specifications

25800 mg/kg LD50, oral (Rat)
50-99-7
C6H12O6
Dextrose
1 S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11 H,1H2/t2-,3-,4+,5-,6+/m1/s1
OCC1OC(O)C(O)C(O)C1O
180.16
250 g
C6H12O6
MFCD00063774
470 g/L
WQZGKKKJIJFFOK-UHFFFAOYNA-N
6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Videos
SDS
Documents
Certificates