missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals α-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%

Catalog Number p-137966
Click to view available options
Quantity:
50 mg
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19046-60-7
C6H11K2O9P
336.32
MFCD00065475
KCIDZIIHRGYJAE-YGFYJFDDSA-L
alpha-d-galactose-1-phosphate dipotassium salt dihydrate
131875454
potassium;[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
[K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
This item is not returnable. View return policy
19046-60-7
C6H11K2O9P
336.32
MFCD00065475
KCIDZIIHRGYJAE-YGFYJFDDSA-L
alpha-d-galactose-1-phosphate dipotassium salt dihydrate
131875454
potassium;[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
[K+].[K+].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to White
CAS Min % 97.5
CAS Max % 100.0
Physical Form Crystalline Powder and/or Crystals
Quantity 50 mg
Assay Percent Range 98%
Solubility Information (5% in water) Clear colorless
Formula Weight 426.39
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Appearance (5% aq. soln.) clear, colorless
Chemical Name or Material α-D-Galactose 1-phosphate dipotassium salt pentahydrate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'