missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Galactosamine Hydrochloride, 99%, ACROS Organics™

Brand:  Acros Organics 160440050

Code : 29

Additional Details : CAS Number : 1772-03-8 Weight : 0.06000kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10153640

  • £140.30 / 5g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

D(+)-Galactosamine hydrochloride
≥98.5%
C6H13NO5·HCl
MFCD00135830
1772-03-8
99%
215.64
Chondrosamine hydrochloride, 2-Amino-2-deoxy-D-galactose hydrochloride

Specifications

D(+)-Galactosamine hydrochloride
1772-03-8
Fine Crystalline Powder
99%
215.64
Chondrosamine hydrochloride, 2-Amino-2-deoxy-D-galactose hydrochloride
Glass bottle
+93° to +99° (20°C, 589nm) (c=1, H2O), after 24 hrs.
+96° (20°C 589nm c=1,H2O,24 hr.)
5g
(10% aq. soln.) clear, colorless
≥98.5%
C6H13NO5·HCl
MFCD00135830
Authentic
1% max.
1% max. (K.F.)
White to off-white
Videos
SDS