missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ D-Galactal, 98%

Brand:  Alfa Aesar™ J66129.03

Code : NEW

Additional Details : CAS Number : 21193-75-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497369

  • £28.20 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

D-Galactal
C6H10O4
MFCD00038067
1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol
21193-75-9
146.14
81690

Specifications

D-Galactal
1g
Powder
C6H10O4
MFCD00038067
1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol
99° to 103°C
−21.5° (c=1.2 in Methanol)
21193-75-9
98%
146.14
81690
White to Off-white
Videos
SDS