missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Cysteine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals
48.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
500 mg
1 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Yellow |
Packaging | Glass Bottle |
Formula Weight | 177.77 |
Quantity | 1 g |
Physical Form | Crystalline Powder |
Assay | 98% |
Chemical Name or Material | D-Cysteine hydrochloride monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'