missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Carvone, 96%, natural, Thermo Scientific Chemicals

Catalog Number p-83524
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2244-16-8
C10H14O
150.22
MFCD00062997
ULDHMXUKGWMISQ-VIFPVBQESA-N
d-carvone, +-carvone, s-+-carvone, s-carvone, s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, +-s-carvone, carvone, +, d +-carvone, carvol, s-+-p-mentha-6,8-dien-2-one
16724
CHEBI:15399
(5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
CC1=CCC(CC1=O)C(=C)C
This item is not returnable. View return policy
2244-16-8
C10H14O
150.22
MFCD00062997
ULDHMXUKGWMISQ-VIFPVBQESA-N
d-carvone, +-carvone, s-+-carvone, s-carvone, s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, +-s-carvone, carvone, +, d +-carvone, carvol, s-+-p-mentha-6,8-dien-2-one
16724
CHEBI:15399
(5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
CC1=CCC(CC1=O)C(=C)C

D(+)-Carvone, CAS # 2244-16-8, is a monoterpene ketone, a natural product found mainly in Achillea ageratum and Curcuma pierreana. It can be used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• D(+)-Carvone belongs to terpene family and is considered a monoterpene ketone naturally found mainly in Achillea ageratum and Curcuma pierreana
Application
• D(+)-Carvone has shown a suppressant effect against high-fat diet induced weight gain in mice
• It has been used as a flavoring agent
• It has been used in organic synthesis as starting material for the asymmetric total synthesis of natural products, including the synthesis of the terpenoid quassin
• In vitro studies demonstrated that this compound can inhibit cellulase-induced capsidiol production in Nicotiana tabacum by interfering with protein isoprenylation
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.9560g/mL
Boiling Point 230.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.5010
Quantity 25 g
Beilstein 07, 153
Merck Index 14, 1874
Specific Gravity 0.956
Specific Rotation + 57.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, chloroform,, propylene glycol and mineral oils, insoluble in glycerol
Formula Weight 150.22
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material D(+)-Carvone, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'