missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Camphoric acid, 99%, Thermo Scientific Chemicals

Catalog Number p-3776525
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
124-83-4
C10H14O4-2
198.218
MFCD00001375
LSPHULWDVZXLIL-LDWIPMOCSA-L
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
6918944
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C
This item is not returnable. View return policy
124-83-4
C10H14O4-2
198.218
MFCD00001375
LSPHULWDVZXLIL-LDWIPMOCSA-L
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
6918944
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 183°C to 187°C
Color White
Assay Percent Range 99%
Beilstein 09, 745
Fieser 11,352; 15,67
Merck Index 15, 1735
Formula Weight 200.23
Percent Purity ≥98.5%
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Camphoric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'