missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-beta-Prolinol, 95%, Thermo Scientific Chemicals
58.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
It is an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Store at room temperature.
It is an intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Store at room temperature.
Specifications
Density | 0.979 |
Boiling Point | 173°C to 177°C |
Quantity | 250 mg |
Sensitivity | Hygroscopic |
Solubility Information | Slightly soluble in water. |
Formula Weight | 101.15 |
Percent Purity | 95% |
Chemical Name or Material | D-beta-Prolinol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'