missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Arabinose (White Powder), Fisher BioReagents

Brand:  Fisher Bioreagents BP2504-25

Code : 32

Additional Details : CAS Number : 28697-53-2 Weight : 0.07500kg

Quantity 25g
Packaging Amber Glass

 View more versions of this product

Product Code. 11433064

  • £118.70 / 25g


Estimated Shipment
22-06-2018
Add to basket

Description

Description

D-(-)-Arabinose is a component used for culture media.
Specifications

Chemical Identifiers

D-(-)-Arabinose
Powder/Solid
C5H10O5
28697-53-2
≥99 %
150.07

Specifications

D-(-)-Arabinose
Conforms to reference
Powder/Solid
C5H10O5
150.07
-107 to -97° (+ or -)
152°C
28697-53-2
Pass Test
≥99 %
Amber Glass
99% min.
White
25g
Videos
SDS Documentation

Technical Documents