missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(-)-Arabinose, 99%

Catalog Number p-7080709
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
10323-20-3
C5H10O5
150.13
MFCD00135608
PYMYPHUHKUWMLA-WDCZJNDASA-N
d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
66308
CHEBI:46983
(2S,3R,4R)-2,3,4,5-tetrahydroxypentanal
C(C(C(C(C=O)O)O)O)O
This item is not returnable. View return policy
10323-20-3
C5H10O5
150.13
MFCD00135608
PYMYPHUHKUWMLA-WDCZJNDASA-N
d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
66308
CHEBI:46983
(2S,3R,4R)-2,3,4,5-tetrahydroxypentanal
C(C(C(C(C=O)O)O)O)O

D-(-)-Arabinose is used in culture media for some bacteria. It serves as a component of biopolymers such as hemicellulose and pectin. In the synthetic biology laboratory, it is employed as a reversible switch for protein expression under the Pbad promoter (i.e. part of plasmids) in E. coli. Further, it acts as an inhibitor of the enzyme glucose dehydrogenase. In addition to this, it is used as a pharmaceutical intermediate and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Arabinose is used in culture media for some bacteria. It serves as a component of biopolymers such as hemicellulose and pectin. In the synthetic biology laboratory, it is employed as a reversible switch for protein expression under the Pbad promoter (i.e. part of plasmids) in E. coli. Further, it acts as an inhibitor of the enzyme glucose dehydrogenase. In addition to this, it is used as a pharmaceutical intermediate and food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 154°C to 158°C
Quantity 10 g
Beilstein 1723079
Solubility Information Soluble in water.
Optical Rotation −104° (c=10 in water, 24h)
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material D- (-)-Arabinose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'