Learn More
D-AP5, Tocris Bioscience™
Potent and selective NMDA antagonist; more active form of DL-AP5
Brand:  Tocris (Bio-Techne) 0106/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13437433
  • £610.05 / 50mg
Estimated Shipment date
from Supplier 25-11-2019
Description
Description
Widely used competitive NMDA antagonist. More active form of AP5. Also agonist at quisqualate-sensitized AP6 site where it is less potent than the L-isomer, L-AP5 (Cat. No.0107).
Available as part of the Mixed NMDA Receptor Tocriset™ . DL Mixture and L-isomer also available.
Specifications
Chemical Identifiers
79055-68-8
197.127
d-ap5, 5-phosphono-d-norvaline, d-norvaline, 5-phosphono, d-apv, 2r-2-amino-5-phosphonopentanoic acid, d-2-amino-5-phosphopentanoic acid, d---2-amino-5-phosphonopentanoic acid, d--apv, d-2-amino-5-phosphonovaleric acid, d--ap-5
(2R)-2-amino-5-phosphonopentanoic acid
C5H12NO5P
VOROEQBFPPIACJ-SCSAIBSYSA-N
135342
C(CC(C(=O)O)N)CP(=O)(O)O
Specifications
50mg
79055-68-8
d-ap5, 5-phosphono-d-norvaline, d-norvaline, 5-phosphono, d-apv, 2r-2-amino-5-phosphonopentanoic acid, d-2-amino-5-phosphopentanoic acid, d---2-amino-5-phosphonopentanoic acid, d--apv, d-2-amino-5-phosphonovaleric acid, d--ap-5
C(CC(C(=O)O)N)CP(=O)(O)O
197.127
197.13
D-AP5
C5H12NO5P
VOROEQBFPPIACJ-SCSAIBSYSA-N
(2R)-2-amino-5-phosphonopentanoic acid
135342
>99%