missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.
Solubility
Soluble in Water 100 mg/ml.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.
Solubility
Soluble in Water 100 mg/ml.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 108°C to 110°C |
Color | White |
Beilstein | 4235975 |
Solubility Information | Soluble in Water 100mg/ml. |
Formula Weight | 139.58 |
Percent Purity | 98% |
Quantity | 5 g |
Physical Form | Crystalline Powder |
Chemical Name or Material | D-Alanine methyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'